(S)-7-Amino-N-(4-fluorobenzyl)-8-oxo-2,9,16,18,21-pentaazabicyclo[15.3.1]henicosa-1(20),16,18-triene-20-carboxamide ≥98%

Supplier: Ambeed
Warning

A1177155-1MG A1177155-5MG A1177155-10MG A1177155-25MG A1177155-50MG
77596-120EA 23.58 USD
77596-120 77620-572 77866-345 77866-346 77866-347
(S)-7-Amino-N-(4-fluorobenzyl)-8-oxo-2,9,16,18,21-pentaazabicyclo[15.3.1]henicosa-1(20),16,18-triene-20-carboxamide ≥98%
(S)-7-Amino-N-(4-fluorobenzyl)-8-oxo-2,9,16,18,21-pentaazabicyclo[15.3.1]henicosa-1(20),16,18-triene-20-carboxamide

Precaución: for Research Use Only

Formula: C₂₄H₃₄FN₇O₂
MDL Number: MFCD32174280
CAS Number: 1612782-86-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR