4-((trans-4-(2-Methoxyethoxy)cyclohexyl)amino)-1-methyl-6-(thiazol-5-yl)quinolin-2(1H)-one ≥98%

Supplier: Ambeed
Warning

A1177137-100MG A1177137-25MG A1177137-50MG A1177137-10MG
77620-566EA 494.4 USD
77620-566 77620-568 77620-570 77866-344
4-((trans-4-(2-Methoxyethoxy)cyclohexyl)amino)-1-methyl-6-(thiazol-5-yl)quinolin-2(1H)-one ≥98%
4-((trans-4-(2-Methoxyethoxy)cyclohexyl)amino)-1-methyl-6-(thiazol-5-yl)quinolin-2(1H)-one

Precaución: for Research Use Only

Formula: C₂₂H₂₇N₃O₃S
MDL Number: MFCD32062812
CAS Number: 1700637-55-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR