4-Ethyl-5-methyl-5,6-dihydro-[1,3]dioxolo[4,5-j]phenanthridine ≥99%

Supplier: Ambeed
Danger

Synonyms: HLY78

A1177124-1MG A1177124-10MG A1177124-5MG
77596-118EA 32.8 USD
77596-118 77620-562 77620-564
4-Ethyl-5-methyl-5,6-dihydro-[1,3]dioxolo[4,5-j]phenanthridine ≥99%
4-Ethyl-5-methyl-5,6-dihydro-[1,3]dioxolo[4,5-j]phenanthridine

Precaución: for Research Use Only

Formula: C₁₇H₁₇NO₂
MW: 267.32 g/mol
MDL Number: MFCD26961120
CAS Number: 854847-61-3
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR