(2-(1H-Indol-3-yl)-1H-imidazol-5-yl)(3,4,5-trimethoxyphenyl)methanone ≥95%

Supplier: Ambeed
Warning

Synonyms: Sabizabulin

A1177099-1MG A1177099-5MG
77596-116EA 104.55 USD
77596-116 77620-560
(2-(1H-Indol-3-yl)-1H-imidazol-5-yl)(3,4,5-trimethoxyphenyl)methanone ≥95%
(2-(1H-Indol-3-yl)-1H-imidazol-5-yl)(3,4,5-trimethoxyphenyl)methanone

Precaución: for Research Use Only

Formula: C₂₁H₁₉N₃O₄
MW: 377.39 g/mol
CAS Number: 1332881-26-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR