Methyl 2-(1-(5-chloro-7H-pyrrolo[2,3-d]pyrimidin-4-yl)piperidin-4-yl)-1H-benzo[d]imidazole-6-carboxylate 98+%

Supplier: Ambeed

A1177091-10MG A1177091-1MG A1177091-5MG
77596-106EA 29.73 USD
77596-106 77596-108 77596-110
Methyl 2-(1-(5-chloro-7H-pyrrolo[2,3-d]pyrimidin-4-yl)piperidin-4-yl)-1H-benzo[d]imidazole-6-carboxylate 98+%
Methyl 2-(1-(5-chloro-7H-pyrrolo[2,3-d]pyrimidin-4-yl)piperidin-4-yl)-1H-benzo[d]imidazole-6-carboxylate

Precaución: for Research Use Only

Formula: C₂₀H₁₉ClN₆O₂
MDL Number: MFCD32067935
CAS Number: 2097938-51-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR