1-Methyl-4-(4-morpholino-7H-pyrrolo[2,3-d]pyrimidin-5-yl)-1H-pyrrole-2-carbonitrile ≥98%

Supplier: Ambeed

Synonyms: PFE-360

A1177088-5MG A1177088-10MG A1177088-1MG
77596-104EA 161.95 USD
77596-104 77596-100 77596-102
1-Methyl-4-(4-morpholino-7H-pyrrolo[2,3-d]pyrimidin-5-yl)-1H-pyrrole-2-carbonitrile ≥98%
1-Methyl-4-(4-morpholino-7H-pyrrolo[2,3-d]pyrimidin-5-yl)-1H-pyrrole-2-carbonitrile

Precaución: for Research Use Only

Formula: C₁₆H₁₆N₆O
MW: 308.34 g/mol
MDL Number: MFCD31697701
CAS Number: 1527475-61-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR