3-(3-(1-Cyclopropyl-1H-pyrazol-4-yl)-1H-indazol-6-yl)-4-methyl-N-(3-(trifluoromethyl)phenyl)benzamide 98+%

Supplier: Ambeed

A1176996-100MG A1176996-250MG
77866-334EA 650 USD
77866-334 77866-335
3-(3-(1-Cyclopropyl-1H-pyrazol-4-yl)-1H-indazol-6-yl)-4-methyl-N-(3-(trifluoromethyl)phenyl)benzamide 98+%
3-(3-(1-Cyclopropyl-1H-pyrazol-4-yl)-1H-indazol-6-yl)-4-methyl-N-(3-(trifluoromethyl)phenyl)benzamide

Precaución: for Research Use Only

Formula: C₂₈H₂₂F₃N₅O
MDL Number: MFCD32067903
CAS Number: 2308497-58-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR