N-(2-((2-(Dimethylamino)ethyl)(methyl)amino)-5-((4-(1-methyl-1H-indol-3-yl)pyrimidin-2-yl)amino)-6-(2,2,2-trifluoroethoxy)pyridin-3-yl)acrylamide methanesulfonate ≥99%
Supplier: Ambeed
|
Warning
|
Synonyms:
Firmonertinib mesylate
A1176940-10MG
A1176940-100MG
A1176940-250MG
A1176940-25MG
A1176940-50MG
77620-546EA
105.06
USD
77620-546
77866-319
77866-320
77866-321
77866-322
N-(2-((2-(Dimethylamino)ethyl)(methyl)amino)-5-((4-(1-methyl-1H-indol-3-yl)pyrimidin-2-yl)amino)-6-(2,2,2-trifluoroethoxy)pyridin-3-yl)acrylamide methanesulfonate ≥99%
N-(2-((2-(Dimethylamino)ethyl)(methyl)amino)-5-((4-(1-methyl-1H-indol-3-yl)pyrimidin-2-yl)amino)-6-(2,2,2-trifluoroethoxy)pyridin-3-yl)acrylamide methanesulfonate
Precaución: for Research Use Only
Formula:
C₂₉H₃₅F₃N₈O₅S MW: 664.7 g/mol |
MDL Number:
MFCD31746903 CAS Number: 2130958-55-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...