(S)-N-(1-((5-(N-(tert-Butyl)sulfamoyl)naphthalen-1-yl)amino)-1-oxo-3-phenylpropan-2-yl)cyclohexanecarboxamide 98.96%

Supplier: Ambeed

A1176859-10MG A1176859-1MG A1176859-5MG
77596-082EA 102.5 USD
77596-082 77596-084 77596-086
(S)-N-(1-((5-(N-(tert-Butyl)sulfamoyl)naphthalen-1-yl)amino)-1-oxo-3-phenylpropan-2-yl)cyclohexanecarboxamide 98.96%
(S)-N-(1-((5-(N-(tert-Butyl)sulfamoyl)naphthalen-1-yl)amino)-1-oxo-3-phenylpropan-2-yl)cyclohexanecarboxamide

Precaución: for Research Use Only

Formula: C₃₀H₃₇N₃O₄S
MDL Number: MFCD31813673
CAS Number: 2241025-68-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR