3-((1S,2S)-1-(5-((S)-2,2-Dimethyltetrahydro-2H-pyran-4-yl)-2-((S)-3-(3-(4-fluoro-1-methyl-1H-indazol-5-yl)-2-oxo-2,3-dihydro-1H-imidazol-1-yl)-2-(4-fluoro-3,5-dimethylphenyl)-4-methyl-4,5,6,7-tetrahydro-2H-pyrazolo[4,3-c]pyridine-5-carbonyl)-1H-indol-1-yl ≥98%

Supplier: Ambeed
Warning

Synonyms: Orforglipron

A1176829-100MG A1176829-10MG A1176829-25MG A1176829-50MG A1176829-5MG A1176829-250MG
77620-528EA 319.3 USD
77620-528 77620-530 77620-532 77620-534 77620-536 77866-312
3-((1S,2S)-1-(5-((S)-2,2-Dimethyltetrahydro-2H-pyran-4-yl)-2-((S)-3-(3-(4-fluoro-1-methyl-1H-indazol-5-yl)-2-oxo-2,3-dihydro-1H-imidazol-1-yl)-2-(4-fluoro-3,5-dimethylphenyl)-4-methyl-4,5,6,7-tetrahydro-2H-pyrazolo[4,3-c]pyridine-5-carbonyl)-1H-indol-1-yl ≥98%
3-((1S,2S)-1-(5-((S)-2,2-Dimethyltetrahydro-2H-pyran-4-yl)-2-((S)-3-(3-(4-fluoro-1-methyl-1H-indazol-5-yl)-2-oxo-2,3-dihydro-1H-imidazol-1-yl)-2-(4-fluoro-3,5-dimethylphenyl)-4-methyl-4,5,6,7-tetrahydro-2H-pyrazolo[4,3-c]pyridine-5-carbonyl)-1H-indol-1-y

Precaución: for Research Use Only

Formula: C₄₈H₄₈F₂N₁₀O₅
MDL Number: MFCD31807602
CAS Number: 2212020-52-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR