1-((1R,3R,4R,7S)-1-((Bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-7-hydroxy-2,5-dioxabicyclo[2.2.1]heptan-3-yl)-5-methylpyrimidine-2,4(1H,3H)-dione ≥98%

Supplier: Ambeed
Warning

Synonyms: Locked nucleic acid 1

A1176777-100MG A1176777-1G A1176777-250MG
77571-062EA 65.92 USD
77571-062 77620-524 77620-526
1-((1R,3R,4R,7S)-1-((Bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-7-hydroxy-2,5-dioxabicyclo[2.2.1]heptan-3-yl)-5-methylpyrimidine-2,4(1H,3H)-dione ≥98%
1-((1R,3R,4R,7S)-1-((Bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-7-hydroxy-2,5-dioxabicyclo[2.2.1]heptan-3-yl)-5-methylpyrimidine-2,4(1H,3H)-dione

Precaución: for Research Use Only

Formula: C₃₂H₃₂N₂O₈
MW: 572.61 g/mol
MDL Number: MFCD31537291
CAS Number: 206055-71-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR