5-Chloro-N-((4-(6-fluoro-7-(methylamino)-2,4-dioxo-1,4-dihydroquinazolin-3(2H)-yl)phenyl)carbamoyl)thiophene-2-sulfonamide ≥98%

Supplier: Ambeed
Danger

Synonyms: Elinogrel

A1176677-10MG A1176677-5MG
77620-516EA 141.11 USD
77620-516 77620-518
5-Chloro-N-((4-(6-fluoro-7-(methylamino)-2,4-dioxo-1,4-dihydroquinazolin-3(2H)-yl)phenyl)carbamoyl)thiophene-2-sulfonamide ≥98%
5-Chloro-N-((4-(6-fluoro-7-(methylamino)-2,4-dioxo-1,4-dihydroquinazolin-3(2H)-yl)phenyl)carbamoyl)thiophene-2-sulfonamide

Precaución: for Research Use Only

Formula: C₂₀H₁₅ClFN₅O₅S₂
MW: 523.95 g/mol
MDL Number: MFCD16619373
CAS Number: 936500-94-6
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR