2-((4-(3,4-Dichlorophenethyl)phenyl)amino)benzoic acid ≥98%

Supplier: Ambeed

Synonyms: PD-118057

A1176636-10MG A1176636-1MG A1176636-5MG
77596-062EA 279.83 USD
77596-062 77596-064 77596-066
2-((4-(3,4-Dichlorophenethyl)phenyl)amino)benzoic acid ≥98%
2-((4-(3,4-Dichlorophenethyl)phenyl)amino)benzoic acid

Precaución: for Research Use Only

Formula: C₂₁H₁₇Cl₂NO₂
MDL Number: MFCD08705328
CAS Number: 313674-97-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR