2-(4-(2-(Trifluoromethyl)phenyl)piperidine-1-carboxamido)benzoic acid 99+%
Supplier: Ambeed
|
Danger
|
Synonyms:
A 1120
A1176615-10MG
A1176615-1MG
A1176615-5MG
77596-056EA
59.45
USD
77596-056
77596-058
77596-060
2-(4-(2-(Trifluoromethyl)phenyl)piperidine-1-carboxamido)benzoic acid 99+%
2-(4-(2-(Trifluoromethyl)phenyl)piperidine-1-carboxamido)benzoic acid
Precaución: for Research Use Only
Formula:
C₂₀H₁₉F₃N₂O₃ MW: 392.3717 g/mol |
MDL Number:
MFCD12912393 CAS Number: 1152782-19-8 UN: 3077 ADR: 9,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...