NU6140 ≥99%

Supplier: Ambeed
Warning

Synonyms: 4-((6-(Cyclohexylmethoxy)-7H-purin-2-yl)amino)-N,N-diethylbenzamide, 4-(6-Cyclohexylmethoxy-9H-purin-2-ylamino)-N,N-diethylbenzamide

A1176612-100MG A1176612-25MG A1176612-50MG
77620-504EA 125.66 USD
77620-504 77620-506 77620-508
NU6140 ≥99%
NU6140

Precaución: for Research Use Only

Formula: C₂₃H₃₀N₆O₂
MW: 422.5 g/mol
MDL Number: MFCD09264153
CAS Number: 444723-13-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR