(R)-3-(Chloro((2-hydroxy-3-(piperidin-1-yl)propoxy)imino)methyl)pyridine 1-oxide maleate ≥97%
Supplier: Ambeed
|
Danger
|
Synonyms:
Arimoclomol maleate
A1176606-1MG
A1176606-100MG
A1176606-10MG
A1176606-25MG
A1176606-50MG
A1176606-5MG
77620-502EA
41.2
USD
77620-502
77866-291
77866-292
77866-293
77866-294
77866-295
(R)-3-(Chloro((2-hydroxy-3-(piperidin-1-yl)propoxy)imino)methyl)pyridine 1-oxide maleate ≥97%
(R)-3-(Chloro((2-hydroxy-3-(piperidin-1-yl)propoxy)imino)methyl)pyridine 1-oxide maleate
Precaución: for Research Use Only
Formula:
C₁₈H₂₄ClN₃O₇ MW: 429.85 g/mol |
MDL Number:
MFCD28143527 CAS Number: 289893-26-1 UN: 2928 ADR: 6.1,8 Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...