6-Amino-5-((R)-1-(2,6-dichloro-3-fluorophenyl)ethoxy)-N-(4-((3S,5R)-3,5-dimethylpiperazine-1-carbonyl)phenyl)pyridazine-3-carboxamide hydrochloride ≥98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Ensartinib 2HCl
A1176592-5MG
A1176592-50MG
A1176592-100MG
A1176592-10MG
A1176592-25MG
77620-500EA
111.24
USD
77620-500
77866-290
77866-287
77866-288
77866-289
6-Amino-5-((R)-1-(2,6-dichloro-3-fluorophenyl)ethoxy)-N-(4-((3S,5R)-3,5-dimethylpiperazine-1-carbonyl)phenyl)pyridazine-3-carboxamide hydrochloride ≥98%
6-Amino-5-((R)-1-(2,6-dichloro-3-fluorophenyl)ethoxy)-N-(4-((3S,5R)-3,5-dimethylpiperazine-1-carbonyl)phenyl)pyridazine-3-carboxamide hydrochloride
Precaución: for Research Use Only
Formula:
C₂₆H₂₈Cl₃FN₆O₃ |
MDL Number:
MFCD31657344 CAS Number: 2137030-98-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...