2-(8-(Benzyloxy)-2-methylimidazo[1,2-a]pyridin-3-yl)acetonitrile ≥98%

Supplier: Ambeed
Warning

Synonyms: SCH28080

A1176583-1MG A1176583-10MG A1176583-5MG
77596-048EA 28.7 USD
77596-048 77620-496 77620-498
2-(8-(Benzyloxy)-2-methylimidazo[1,2-a]pyridin-3-yl)acetonitrile ≥98%
2-(8-(Benzyloxy)-2-methylimidazo[1,2-a]pyridin-3-yl)acetonitrile

Precaución: for Research Use Only

Formula: C₁₇H₁₅N₃O
MW: 277.32 g/mol
MDL Number: MFCD00834620
CAS Number: 76081-98-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR