2-(4-Bromo-2,6-dimethylphenoxy)acetic acid ≥95%

Supplier: Ambeed
Warning

A117637-250MG A117637-1G A117637-100MG
77670-652EA 59 USD
77670-652 77880-950 77880-949
2-(4-Bromo-2,6-dimethylphenoxy)acetic acid ≥95%
2-(4-Bromo-2,6-dimethylphenoxy)acetic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrO₃
MDL Number: MFCD03537971
CAS Number: 455887-97-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR