Zinc 2-methylimidazole Zn 28-30% w/w

Supplier: Ambeed
Danger

Synonyms: 2-Methylimidazole zinc salt

A1176337-1G A1176337-5G A1176337-100MG A1176337-250MG
77592-296EA 71.75 USD
77592-296 77596-038 77620-490 77620-492
Zinc 2-methylimidazole Zn 28-30% w/w
Zinc 2-methylimidazole

Precaución: for Research Use Only

Formula: C₈H₁₀N₄Zn
MW: 227.6 g/mol
MDL Number: MFCD10567011
CAS Number: 59061-53-9
UN: 3263
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR