Potassium 3-Ethoxy-2-methyl-3-oxopropanoate ≥97%

Supplier: Ambeed
Warning

A1175954-5G A1175954-250MG
77592-294EA 305.45 USD
77592-294 77620-484
Potassium 3-Ethoxy-2-methyl-3-oxopropanoate ≥97%
Potassium 3-Ethoxy-2-methyl-3-oxopropanoate

Precaución: for Research Use Only

Formula: C₆H₉KO₄
MW: 184.2316 g/mol
MDL Number: MFCD30532430
CAS Number: 103362-70-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR