Chloro(tetrahydrothiophene)gold(I) ≥98%

Supplier: Ambeed
Danger

A1175869-100MG A1175869-250MG
77571-054EA 68 USD
77571-054 77866-269
Chloro(tetrahydrothiophene)gold(I) ≥98%
Chloro(tetrahydrothiophene)gold(I)

Precaución: for Research Use Only

Formula: C₄H₈AuClS
MDL Number: MFCD28411665
CAS Number: 39929-21-0
UN: 1759
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR