Tris(dibenzylideneacetone)platinum(0) ≥98%

Supplier: Ambeed
Warning

A1175866-100MG A1175866-1G
77571-052EA 54.59 USD
77571-052 77620-480
Tris(dibenzylideneacetone)platinum(0) ≥98%
Tris(dibenzylideneacetone)platinum(0)

Precaución: for Research Use Only

Formula: C₅₁H₄₂O₃Pt
MDL Number: MFCD22666053
CAS Number: 11072-92-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR