Bis(acetonitrile)palladium(II) p-toluenesulfonate ≥98%

Supplier: Ambeed
Danger

A1175833-100MG A1175833-1G A1175833-250MG
77620-474EA 84.46 USD
77620-474 77620-476 77866-268
Bis(acetonitrile)palladium(II) p-toluenesulfonate ≥98%
Bis(acetonitrile)palladium(II) p-toluenesulfonate

Precaución: for Research Use Only

Formula: C₁₈H₂₀N₂O₆PdS₂
MDL Number: MFCD28167054
CAS Number: 114757-66-3
UN: 3439
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR