TAS4464 ≥95%

Supplier: Ambeed
Warning

Synonyms: 7-[5-[(Aminosulphonyl)amino]-5-deoxy-β-D-ribofuranosyl]-5-[2-(2-ethoxy-6-fluorophenyl)ethynyl]-7H-Pyrrolo[2,3-d]pyrimidin-4-aminesyl]-5-[2-(2-ethoxy-6-fluorophenyl)ethynyl]-7H-Pyrrolo[2,3-d]pyrimidin-4-amine

A1175747-25MG A1175747-50MG
77571-048EA 194.67 USD
77571-048 77620-466
TAS4464 ≥95%
TAS4464

Precaución: for Research Use Only

Formula: C₂₁H₂₃FN₆O₆S
MW: 506.51 g/mol
MDL Number: MFCD32067967
CAS Number: 1848959-10-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR