(R)-N-((S)-1-([1,1'-Biphenyl]-2-yl)-2-(diphenylphosphanyl)ethyl)-2-methylpropane-2-sulfinamide ≥98%

Supplier: Ambeed
Warning

A1175261-100MG A1175261-250MG A1175261-50MG
77596-030EA 148.63 USD
77596-030 77620-456 77620-458
(R)-N-((S)-1-([1,1'-Biphenyl]-2-yl)-2-(diphenylphosphanyl)ethyl)-2-methylpropane-2-sulfinamide ≥98%
(R)-N-((S)-1-([1,1'-Biphenyl]-2-yl)-2-(diphenylphosphanyl)ethyl)-2-methylpropane-2-sulfinamide

Precaución: for Research Use Only

Formula: C₃₀H₃₂NOPS
MDL Number: MFCD32697169
CAS Number: 1936438-14-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR