Methanesulfonato(2-bis(3,5-di(trifluoromethyl)phenylphosphino)-3,6-dimethoxy-2',6'-bis(dimethylamino)-1,1'-biphenyl )(2'-methylamino-1,1'-biphenyl-2-yl)palladium(II) ≥98%

Supplier: Ambeed
Warning

A1175169-100MG A1175169-250MG
77571-044EA 147 USD
77571-044 77620-434
Methanesulfonato(2-bis(3,5-di(trifluoromethyl)phenylphosphino)-3,6-dimethoxy-2',6'-bis(dimethylamino)-1,1'-biphenyl )(2'-methylamino-1,1'-biphenyl-2-yl)palladium(II) ≥98%
Methanesulfonato(2-bis(3,5-di(trifluoromethyl)phenylphosphino)-3,6-dimethoxy-2',6'-bis(dimethylamino)-1,1'-biphenyl )(2'-methylamino-1,1'-biphenyl-2-yl)palladium(II)

Precaución: for Research Use Only

Formula: C₄₈H₄₄F₁₂N₃O₅PPdS
CAS Number: 1810068-35-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR