N-(2',4'-Difluoro-5-(5-(1-methyl-1H-pyrazol-4-yl)-1H-benzo[d]imidazol-1-yl)-[1,1'-biphenyl]-3-yl)cyclopropanesulfonamide 99+%

Supplier: Ambeed
Warning

A1173509-100MG A1173509-10MG A1173509-1MG A1173509-50MG
77620-416EA 554.23 USD
77620-416 77620-418 77620-420 77866-243
N-(2',4'-Difluoro-5-(5-(1-methyl-1H-pyrazol-4-yl)-1H-benzo[d]imidazol-1-yl)-[1,1'-biphenyl]-3-yl)cyclopropanesulfonamide 99+%
N-(2',4'-Difluoro-5-(5-(1-methyl-1H-pyrazol-4-yl)-1H-benzo[d]imidazol-1-yl)-[1,1'-biphenyl]-3-yl)cyclopropanesulfonamide

Precaución: for Research Use Only

Formula: C₂₆H₂₁F₂N₅O₂S
MDL Number: MFCD32062783
CAS Number: 1430723-35-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR