MitoTEMPO ≥98%

Supplier: Ambeed
Warning

A1173173-10MG A1173173-5MG
77620-412EA 74.16 USD
77620-412 77620-414
MitoTEMPO ≥98%
MitoTEMPO

Precaución: for Research Use Only

Formula: C₂₉H₃₅ClN₂O₂P
MW: 510.03 g/mol
MDL Number: MFCD26406410
CAS Number: 1334850-99-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR