N-(2,3-Dihydrobenzo[b][1,4]dioxin-6-yl)-1,4-dihydroindeno[1,2-c]pyrazol-3-amine ≥99%

Supplier: Ambeed
Warning

Synonyms: GN44028

A1172999-1MG A1172999-5MG
77596-022EA 102.5 USD
77596-022 77620-408
N-(2,3-Dihydrobenzo[b][1,4]dioxin-6-yl)-1,4-dihydroindeno[1,2-c]pyrazol-3-amine ≥99%
N-(2,3-Dihydrobenzo[b][1,4]dioxin-6-yl)-1,4-dihydroindeno[1,2-c]pyrazol-3-amine

Precaución: for Research Use Only

Formula: C₁₈H₁₅N₃O₂
MW: 305.33 g/mol
MDL Number: MFCD30182340
CAS Number: 1421448-26-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR