Boc-Met(O)-Oh ≥96%

Supplier: Ambeed
Warning

Synonyms: Boc-Met(O)-OH, N-Boc-L-methionine sulphoxide

A117134-1G A117134-5G
77599-694EA 13.33 USD
77599-694 77670-648
Boc-Met(O)-Oh ≥96%
Boc-Met(O)-Oh

Precaución: for Research Use Only

Formula: C₁₀H₁₉NO₅S
MW: 265.324 g/mol
MDL Number: MFCD00037172
CAS Number: 34805-21-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR