Benzyl-4-bromobutyl ether ≥97%

Supplier: Ambeed
Warning

Synonyms: Benzyl 4-bromobutyl ether, (4-Bromobutoxymethyl)benzene, ((4-Bromobutoxy)methyl)benzene, 4-Benzyloxybutyl Bromide

A116869-1G A116869-5G
77596-018EA 37.92 USD
77596-018 77876-860
Benzyl-4-bromobutyl ether ≥97%
Benzyl-4-bromobutyl ether

Precaución: for Research Use Only

Formula: C₁₁H₁₅BrO
MDL Number: MFCD00155128
CAS Number: 60789-54-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR