Dichlorobis(tricyclohexylphosphine)cobalt(II) ≥98%
Supplier: Ambeed
|
Warning
|
A1168420-100MG
A1168420-1G
A1168420-250MG
77596-008EA
56.38
USD
77596-008
77596-010
77596-012
Dichlorobis(tricyclohexylphosphine)cobalt(II) ≥98%
Dichlorobis(tricyclohexylphosphine)cobalt(II)
Precaución: for Research Use Only
Formula:
C₃₆H₆₈Cl₂CoP₂ MW: 692.71 g/mol |
MDL Number:
MFCD31807340 CAS Number: 14726-62-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...