Bis[(tetrabutylammonium iodide)copper(I) iodide] ≥98%

Supplier: Ambeed
Warning

A1168410-1G A1168410-250MG A1168410-5G A1168410-100MG
77571-020EA 223 USD
77571-020 77596-004 77596-006 77866-232
Bis[(tetrabutylammonium iodide)copper(I) iodide] ≥98%
Bis[(tetrabutylammonium iodide)copper(I) iodide]

Precaución: for Research Use Only

Formula: C₁₆H₃₆CuI₂N
MDL Number: MFCD22200557
CAS Number: 81412-07-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR