5-Bromo-4-fluoro-2-iodoaniline ≥97%

Supplier: Ambeed
Warning

A1168216-1G A1168216-250MG
77866-226EA 54 USD
77866-226 77866-227
5-Bromo-4-fluoro-2-iodoaniline ≥97%
5-Bromo-4-fluoro-2-iodoaniline

Precaución: for Research Use Only

Formula: C₆H₄BrFIN
MDL Number: MFCD30834158
CAS Number: 1265236-37-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR