(1R,2R)-2-((tert-Butoxycarbonyl)amino)cyclohexanecarboxylic acid ≥97%

Supplier: Ambeed
Warning

A1167939-1G A1167939-250MG
77592-290EA 774.9 USD
77592-290 77620-400
(1R,2R)-2-((tert-Butoxycarbonyl)amino)cyclohexanecarboxylic acid ≥97%
(1R,2R)-2-((tert-Butoxycarbonyl)amino)cyclohexanecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₂H₂₁NO₄
MW: 243.2994 g/mol
MDL Number: MFCD04974225
CAS Number: 233661-54-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR