3-Cyano-N-(3-(1-isobutyrylpiperidin-4-yl)-1-methyl-4-(trifluoromethyl)-1H-pyrrolo[2,3-b]pyridin-5-yl)benzamide ≥98%

Supplier: Ambeed
Warning

Synonyms: PF-06747711

A1167892-100MG A1167892-25MG A1167892-50MG
77866-222EA 768 USD
77866-222 77866-223 77866-224
3-Cyano-N-(3-(1-isobutyrylpiperidin-4-yl)-1-methyl-4-(trifluoromethyl)-1H-pyrrolo[2,3-b]pyridin-5-yl)benzamide ≥98%
3-Cyano-N-(3-(1-isobutyrylpiperidin-4-yl)-1-methyl-4-(trifluoromethyl)-1H-pyrrolo[2,3-b]pyridin-5-yl)benzamide

Precaución: for Research Use Only

Formula: C₂₆H₂₆F₃N₅O₂
MDL Number: MFCD31697703
CAS Number: 1892576-58-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR