CITCO ≥98%

Supplier: Ambeed
Warning

Synonyms: 6-(4-Chlorophenyl)imidazo[2,1-b]thiazole-5-carbaldehyde O-(3,4-dichlorobenzyl) oxime, 6-(4-Chlorophenyl)imidazo[2,1-b] [1,3]thiazole-5-carbaldehyde O-3,4-dichlorobenzyl) oxime

A1167756-100MG A1167756-250MG A1167756-25MG A1167756-50MG
77595-998EA 205 USD
77595-998 77620-392 77620-394 77620-396
CITCO ≥98%
CITCO

Precaución: for Research Use Only

Formula: C₁₉H₁₂Cl₃N₃OS
MW: 436.7 g/mol
MDL Number: MFCD00139608
CAS Number: 338404-52-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR