(3S,6S,9S,12R,15S,18S,21S,24S,30S,33S)-30-Ethyl-33-((1R,2R,E)-1-hydroxy-2-methylhepta-4,6-dien-1-yl)-6,9,18,24-tetraisobutyl-3,21-diisopropyl-1,4,7,10,12,15,19,25,28-nonamethyl-1,4,7,10,13,16,19,22,25,28,31-undecaazacyclotritriacontan-2,5,8,11,14,17,20,23,26,29,32-undecaone ≥98%

Supplier: Ambeed
Danger

A1167646-1MG A1167646-10MG A1167646-5MG
77595-996EA 28.7 USD
77595-996 77620-388 77620-390
(3S,6S,9S,12R,15S,18S,21S,24S,30S,33S)-30-Ethyl-33-((1R,2R,E)-1-hydroxy-2-methylhepta-4,6-dien-1-yl)-6,9,18,24-tetraisobutyl-3,21-diisopropyl-1,4,7,10,12,15,19,25,28-nonamethyl-1,4,7,10,13,16,19,22,25,28,31-undecaazacyclotritriacontan-2,5,8,11,14,17,20,23,26,29,32-undecaone ≥98%
(3S,6S,9S,12R,15S,18S,21S,24S,30S,33S)-30-Ethyl-33-((1R,2R,E)-1-hydroxy-2-methylhepta-4,6-dien-1-yl)-6,9,18,24-tetraisobutyl-3,21-diisopropyl-1,4,7,10,12,15,19,25,28-nonamethyl-1,4,7,10,13,16,19,22,25,28,31-undecaazacyclotritriacontan-2,5,8,11,14,17,20,2

Precaución: for Research Use Only

Formula: C₆₃H₁₁₁N₁₁O₁₂
MW: 1214.62 g/mol
MDL Number: MFCD30720011
CAS Number: 515814-01-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR