1-(4-Benzylpiperidin-1-yl)-3-(3,5-dimethyl-1-(3-methyl-[1,2,4]triazolo[4,3-b]pyridazin-6-yl)-1H-pyrazol-4-yl)propan-1-one ≥98%

Supplier: Ambeed
Warning

Synonyms: C25-140

A1167134-1MG A1167134-10MG A1167134-5MG
77595-990EA 32.8 USD
77595-990 77620-374 77620-376
1-(4-Benzylpiperidin-1-yl)-3-(3,5-dimethyl-1-(3-methyl-[1,2,4]triazolo[4,3-b]pyridazin-6-yl)-1H-pyrazol-4-yl)propan-1-one ≥98%
1-(4-Benzylpiperidin-1-yl)-3-(3,5-dimethyl-1-(3-methyl-[1,2,4]triazolo[4,3-b]pyridazin-6-yl)-1H-pyrazol-4-yl)propan-1-one

Precaución: for Research Use Only

Formula: C₂₆H₃₁N₇O
MW: 457.57 g/mol
MDL Number: MFCD27163630
CAS Number: 1358099-18-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR