2,6-Bis(N-pyrazolyl)pyridine nickel (II) dichloride ≥98%

Supplier: Ambeed
Danger

A1167067-1G A1167067-250MG A1167067-100MG
77620-370EA 609.76 USD
77620-370 77620-372 77866-210
2,6-Bis(N-pyrazolyl)pyridine nickel (II) dichloride ≥98%
2,6-Bis(N-pyrazolyl)pyridine nickel (II) dichloride

Precaución: for Research Use Only

MDL Number: MFCD32062476
CAS Number: 2304667-33-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR