(2,9-Dimethyl-4,7-diphenyl-1,10-phenanthroline)nickel dibromide ≥98%

Supplier: Ambeed
Danger

A1167056-1G A1167056-5G
77592-288EA 144.53 USD
77592-288 77620-368
(2,9-Dimethyl-4,7-diphenyl-1,10-phenanthroline)nickel dibromide ≥98%
(2,9-Dimethyl-4,7-diphenyl-1,10-phenanthroline)nickel dibromide

Precaución: for Research Use Only

MDL Number: MFCD31807323
CAS Number: 326822-02-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR