PRL-3 Inhibitor ≥98%

Supplier: Ambeed
Danger

Synonyms: PTP4A3 Inhibitor, 5-(5-Bromo-2-((2-bromobenzyl)oxy)benzylidene)-2-thioxothiazolidin-4-one, P0108, BR-1, PRL-3 Inhibitor I, 5-[[5-Bromo-2-[(2-bromophenyl)methoxy]phenyl]methylene]-2-thioxo-4-thiazolidinone

A1166385-100MG A1166385-50MG
77620-364EA 175.1 USD
77620-364 77620-366
PRL-3 Inhibitor ≥98%
PRL-3 Inhibitor

Precaución: for Research Use Only

Formula: C₁₇H₁₁Br₂NO₂S₂
MW: 485.2 g/mol
MDL Number: MFCD11045303
CAS Number: 893449-38-2
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR