Sorbitan monostearate Saponification Value 145-160

Supplier: Ambeed
Warning

Synonyms: Sorbitan monooctadecanoate, (2R,3R,4R,5S)-Hexane-1,2,3,4,5,6-hexaol monooctadecanoate, Sorbitan stearate

A1165384-1000G A1165384-100G
77620-352EA 17.51 USD
77620-352 77620-354
Sorbitan monostearate Saponification Value 145-160
Sorbitan monostearate

Precaución: for Research Use Only

Formula: C₂₄H₄₆O₆
MW: 430.63 g/mol
MDL Number: MFCD00005366
CAS Number: 1338-41-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR