5,5-Difluoro-2,8-diiodo-1,3,7,9-tetramethyl-10-phenyl-5H-dipyrrolo[1,2-c:2',1'-f][1,3,2]diazaborinin-4-ium-5-uide ≥98%

Supplier: Ambeed
Warning

A1164708-1G A1164708-100MG A1164708-250MG
77620-346EA 688.04 USD
77620-346 77866-177 77866-178
5,5-Difluoro-2,8-diiodo-1,3,7,9-tetramethyl-10-phenyl-5H-dipyrrolo[1,2-c:2',1'-f][1,3,2]diazaborinin-4-ium-5-uide ≥98%
5,5-Difluoro-2,8-diiodo-1,3,7,9-tetramethyl-10-phenyl-5H-dipyrrolo[1,2-c:2',1'-f][1,3,2]diazaborinin-4-ium-5-uide

Precaución: for Research Use Only

Formula: C₁₉H₁₇BF₂I₂N₂
MW: 575.9684 g/mol
CAS Number: 1083009-44-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR