1-Chloronaphthalene-5-boronic acid ≥98%

Supplier: Ambeed
Warning

A1163137-1G A1163137-250MG A1163137-100MG A1163137-5G
77620-318EA 466.59 USD
77620-318 77620-320 77866-154 77866-155
1-Chloronaphthalene-5-boronic acid ≥98%
1-Chloronaphthalene-5-boronic acid

Precaución: for Research Use Only

Formula: C₁₀H₈BClO₂
MDL Number: MFCD18411296
CAS Number: 2142655-40-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR