(S)-tert-Butyl 3-(aminomethyl)piperidine-1-carboxylate hydrochloride ≥95%

Supplier: Ambeed
Warning

A116222-1G A116222-250MG A116222-100MG
77610-948EA 145.23 USD
77610-948 77610-950 77880-939
(S)-tert-Butyl 3-(aminomethyl)piperidine-1-carboxylate hydrochloride ≥95%
(S)-tert-Butyl 3-(aminomethyl)piperidine-1-carboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₂₃ClN₂O₂
MW: 250.76 g/mol
MDL Number: MFCD11112271
CAS Number: 1217702-57-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR