ε-Poly-L-lysine 97%, Technical Grade

Supplier: Ambeed
Warning

A116127-25G A116127-500G
77570-986EA 40.17 USD
77570-986 77570-988
ε-Poly-L-lysine 97%, Technical Grade
ε-Poly-L-lysine

Precaución: for Research Use Only

Formula: (C₆H₁₂N₂O)n
MDL Number: MFCD01756704
CAS Number: 28211-04-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR