Methyl-PEG2000-DSPE, M.W. 2000 ≥98%

Supplier: Ambeed
Warning

Synonyms: Poly(oxy-1,2-ethanediyl), α-[6-hydroxy-6-oxido-1,12-dioxo-9-[(1-oxooctadecyl)oxy]-5,7,11-trioxa-2-aza-6-phosphanonacos-1-yl]-ω-methoxy-, 1,2-Distearoyl-phosphatidylethanolamine-methyl-polyethyleneglycol conjugate, DSPE-mPEG (MW 2000), mPEG-DSPE, MW 2000

A1159223-1G A1159223-100MG A1159223-250MG
77592-278EA 80.98 USD
77592-278 77595-944 77595-946
Methyl-PEG2000-DSPE, M.W. 2000 ≥98%
Methyl-PEG2000-DSPE

Precaución: for Research Use Only

MDL Number: MFCD00240077
CAS Number: 147867-65-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR