Ruthenium(2+), bis(2,2′-bipyridine-κN1,κN1′)(dipyrido[3,2-a:2′,3′-c]phenazine-κN4,κN5)-, dichloride ≥97%

Supplier: Ambeed
Warning

A1158991-250MG A1158991-1G
77620-288EA 147.29 USD
77620-288 77866-123
Ruthenium(2+), bis(2,2′-bipyridine-κN1,κN1′)(dipyrido[3,2-a:2′,3′-c]phenazine-κN4,κN5)-, dichloride ≥97%
Ruthenium(2+), bis(2,2′-bipyridine-κN1,κN1′)(dipyrido[3,2-a:2′,3′-c]phenazine-κN4,κN5)-, dichloride

Precaución: for Research Use Only

Formula: C₃₈H₂₆Cl₂N₈Ru
MW: 766.6426 g/mol
CAS Number: 155659-28-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR